Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesityl Oxide, SPEX CertiPrep™
SDP

Catalog No. s2368
Click to view available options
Quantity:
1 mL

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Mesityl Oxide141-79-71000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 141-79-7, 67-56-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
InChI Key SHOJXDKTYKFBRD-UHFFFAOYSA-N
PubChem CID 8858
IUPAC Name 4-methylpent-3-en-2-one
SMILES CC(=CC(=O)C)C

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Chemical Name or Material Mesityl Oxide
Hazard Category Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.