Learn More
Mesitylene-2-sulfonyl chloride, 99%
Description
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis
Solubility
Soluble in toluene (almost transparency).
Notes
Moisture Sensitive. Store under inert gas. Incompatible with strong bases. Store away from moisture.

Specifications
Specifications
Melting Point | 53°C to 57°C |
Boiling Point | 150°C (20 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Irritating |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 1107601 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in toluene (almost transparency). |
Formula Weight | 218.7 |
Show More |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 212-257-8 |
RTECSNumber | DB8930000 |
TSCA | No |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.