Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylacetic acid, 99%

Catalog No. AC126020010
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle

CAS: 104-01-8 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-01-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004345
InChI Key NRPFNQUDKRYCNX-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetic acid, homoanisic acid, 2-4-methoxyphenyl acetic acid, 4-methoxybenzeneacetic acid, benzeneacetic acid, 4-methoxy, 4-methoxyphenyl acetic acid, p-methoxyphenylacetic acid, 2-p-anisyl acetic acid, p-methoxyphenyl acetic acid
PubChem CID 7690
ChEBI CHEBI:55501
IUPAC Name 2-(4-methoxyphenyl)acetic acid
SMILES COC1=CC=C(C=C1)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 84.0°C to 88.0°C
Color White to Yellow
Boiling Point 140.0°C (3.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3OC6H4CH2CO2H
Quantity 1 kg
Beilstein 10, 190
Solubility Information Solubility in water: 6g/L (20 c)
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Methoxyphenylacetic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
EINECSNumber 203-166-4
RTECSNumber AI8960000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.