Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesitylene, 99%, Extra Pure

Catalog No. p-3736822
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Catalog No. AC125582500

Supplier: Thermo Scientific Chemicals 125582500

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 108-67-8 | C9H12 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-67-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00008538
InChI Key AUHZEENZYGFFBQ-UHFFFAOYSA-N
Synonym mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene
PubChem CID 7947
ChEBI CHEBI:34833
IUPAC Name 1,3,5-trimethylbenzene
SMILES CC1=CC(=CC(=C1)C)C

Specifications

Melting Point -45.0°C
Density 0.8675g/mL
Boiling Point 163.0°C to 166.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4980 to 1.5000
Linear Formula C6H3(CH3)3
Quantity 250 mL
Beilstein 05, 406
Specific Gravity 0.8675
Merck Index 15, 5977
Solubility Information Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with alcohol,benzene and ether
Viscosity 0.834 mm2/s (20°C)
Formula Weight 120.19
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Mesitylene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.<br
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 203-604-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.