Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mesitylenesulfonic acid sodium salt hemihydrate, 98%

Click to view available options
Quantity:
25g
100g
500g
6148-75-0
C9H11NaO3S
222.234
MFCD00067712
AOJUNZYQOYSGHT-UHFFFAOYSA-M
sodium mesitylenesulfonate,benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt,sodium 2,4,6-trimethylbenzenesulfonate,sodium 2,4,6-trimethylbenzenesulphonate,mesitylenesulfonic acid sodium salt hemihydrate,mesitylenesulfonic acid sodium salt,benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt 1:1,2,4,6-trimethylbenzenesulfonic acid sodium salt,mesitylene sodium sulfonate,acmc-1bbae
23661859
sodium;2,4,6-trimethylbenzenesulfonate
CC1=CC(=C(C(=C1)C)S(=O)(=O)[O-])C.[Na+]
6148-75-0
C9H11NaO3S
222.234
MFCD00067712
AOJUNZYQOYSGHT-UHFFFAOYSA-M
sodium mesitylenesulfonate,benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt,sodium 2,4,6-trimethylbenzenesulfonate,sodium 2,4,6-trimethylbenzenesulphonate,mesitylenesulfonic acid sodium salt hemihydrate,mesitylenesulfonic acid sodium salt,benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt 1:1,2,4,6-trimethylbenzenesulfonic acid sodium salt,mesitylene sodium sulfonate,acmc-1bbae
23661859
sodium;2,4,6-trimethylbenzenesulfonate
CC1=CC(=C(C(=C1)C)S(=O)(=O)[O-])C.[Na+]
Quantity 25g
Melting Point >300°C
Beilstein 5784990
Formula Weight 231.26 (222.25 Anhydrous)
Percent Purity 98%
Chemical Name or Material Mesitylenesulfonic acid sodium salt hemihydrate