Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso-1,2-bis(1-naphthyl)ethylenediamine 99%, Thermo Scientific™

Catalog No. AC375301000
Click to view available options
Quantity:
100 mg
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117106-39-5
Molecular Formula C22H20N2
Molecular Weight (g/mol) 312.416
MDL Number MFCD05150370
InChI Key VVJVMUUCGDSHBT-SZPZYZBQSA-N
Synonym 1r,2s-1,2-di naphthalen-1-yl ethane-1,2-diamine, 1r,2s-1,2-dinaphthalen-1-ylethane-1,2-diamine, 1r,2s-1,2-bis 1-naphthyl-1,2-ethanediamine
PubChem CID 2734857
IUPAC Name (1R,2S)-1,2-dinaphthalen-1-ylethane-1,2-diamine
SMILES C1=CC=C2C(=C1)C=CC=C2C(C(C3=CC=CC4=CC=CC=C43)N)N

Specifications

Melting Point 174°C to 177°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 100 mg
Formula Weight 312.41
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material meso-1, 2-Bis(1-naphthyl)ethylenediamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.