Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-1,2-Diphenyl-1,2-ethanediol, 95%

Catalog No. ac247680010
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 579-43-1 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064253
InChI Key IHPDTPWNFBQHEB-UHFFFAOYNA-N
Synonym meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
PubChem CID 853018
ChEBI CHEBI:50015
IUPAC Name (1R,2S)-1,2-diphenylethane-1,2-diol
SMILES OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 136.0°C to 137.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(OH)CH(OH)C6H5
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 214.26
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material meso-1, 2-Diphenyl-1, 2-ethanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.