Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific Chemicals

Catalog No. AC147570025
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-36-6
Molecular Formula C4H2Br2O4
Molecular Weight (g/mol) 273.87
MDL Number MFCD00066439
InChI Key FJWGRXKOBIVTFA-XIXRPRMCSA-L
Synonym 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
PubChem CID 641611
IUPAC Name (2S,3R)-2,3-dibromobutanedioic acid
SMILES [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O

Specifications

Melting Point >275.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula HO2CCH(Br)CH(Br)CO2H
Quantity 2.5 kg
Beilstein 02,625
Merck Index 14,3029
Solubility Information Solubility in water: 20g/L (17°C). Other solubilities: soluble in alcohol and ether, sparingly soluble in chloroform
Formula Weight 275.88
Percent Purity 98%
Physical Form Powder
Chemical Name or Material meso-2,3-Dibromosuccinic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.