Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-Butane-1,2,3,4-tetracarboxylic Dianhydride 96.0+%, TCI America™
SDP

Catalog No. B177025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B177025G 25 g
B17705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B177025G Supplier TCI America Supplier No. B177025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17309-39-6
Molecular Formula C8H6O6
Molecular Weight (g/mol) 198.13
MDL Number MFCD00671500
InChI Key OLQWMCSSZKNOLQ-WUCPZUCCSA-N
PubChem CID 54156817
IUPAC Name 3-[(3S)-2,5-dioxooxolan-3-yl]oxolane-2,5-dione
SMILES C1C(C(=O)OC1=O)C2CC(=O)OC2=O

Specifications

Color White
Quantity 25 g
Formula Weight 198.13
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material meso-Butane-1,2,3,4-tetracarboxylic Dianhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.