Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-Erythritol, Spectrum™ Chemical
SDP

Catalog No. 18620740 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-740 5 g
18-620-741 25 g
18-620-742 100 g
18-620-743 500 g
18-620-744 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-740 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ER1235GM
Add to Cart
Add to Cart

Synonym: Phycitol, Erythrit, Erythritol, i-Erythritol, Mesoerythritol

Chemical Identifiers

CAS 149-32-6

Specifications

Boiling Point 329°C to 331°C (624°F to 628°F)
Packaging Bottle
Quantity 5 g
Molecular Weight (g/mol) 122.12
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material meso-Erythritol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.