Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesothelin Antibody (420411), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB32652APCCY7
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Flow CytometrySpecifications
Mesothelin | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
420411 | |
Flow Cytometry | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
Mouse myeloma cell line NS0-derived recombinant human Mesothelin, Glu296-Gly580, Accession # AAH09272 | |
0.1 mL | |
Cancer | |
10232 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction