Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesothelin Antibody (MB-G10), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11085538MFV500
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Flow CytometrySpecifications
Mesothelin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MB-G10 | |
Flow Cytometry | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
This Mesothelin Antibody (MB-G10) was produced in mesothelin-deficient mice by immunizations with plasmid cDNA encoding human Mesothelin full length protein followed by a single boost of a recombinant human mesothelin-Fc fusion protein. (Uniprot: Q13421) | |
0.1 mL | |
Cancer | |
10232 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction