Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesothelin Antibody (MSLN/2131), Janelia Fluor™ 549, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP279858JF549

Catalog No. NB011223


Only null left
Add to Cart

Description

Description

Mesothelin Monoclonal specifically detects Mesothelin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Mesothelin
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
MSLN
Recombinant fragment (around aa 273-407) of human Mesothelin (MSLN) protein (exact sequence is proprietary) (Uniprot: Q13421)
0.1 mL
Cancer
Human, Mouse, Rat
IgG2b κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
MSLN/2131
Immunohistochemistry, Immunohistochemistry-Paraffin
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.