Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesothelin Antibody (rMSLN/8764), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32043220UG
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Mesothelin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
Recombinant fragment (around aa 273-407) of human Mesothelin protein (exact sequence is proprietary) | |
20 μg | |
Cancer | |
10232 | |
Store at 4&drg;C. Do not freeze. | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
rMSLN/8764 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction