Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesothelin Antibody (YP158), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275255
Description
Mesothelin Monoclonal specifically detects Mesothelin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Mesothelin | |
Monoclonal | |
Unconjugated | |
Tissue culture supernatant with 0.05% Sodium Azide | |
MSLN | |
Synthetic peptide corresponding to C-terminus of human mesothelin protein. | |
0.1 mL | |
10232.0 | |
Store at 4°C. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
YP158 | |
Immunohistochemistry 1:10 - 1:15, Immunohistochemistry-Paraffin | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
Rabbit | |
Tissue culture supernatant | |
Primary | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction