Learn More
Mesothelin Monoclonal Antibody (MB-G10), Rockland™

Mouse Monoclonal Antibody
Supplier: Rockland Immunochemicals 200301A87
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is directed against human mesothelin protein. Cross reactivity with homologues from other sources has not been tested.
Mesothelin is a GPI-linked cell-surface glycoprotein, that is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.Specifications
| Mesothelin | |
| Monoclonal | |
| 1 mg/mL | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| Q13421 | |
| MSLN | |
| This antibody was produced in mesothelin-deficient mice by immunizations with plasmid cDNA encoding human MSLN full length protein followed by a single boost of a recombinant human mesothelin-Fc fusion protein. | |
| RUO | |
| 10232 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| MB-G10 | |
| Unconjugated | |
| MSLN | |
| CAK1; CAK1 antigen; megakaryocyte potentiating factor; Megakaryocyte-potentiating factor; Mes; Mesothelin; Mesothelin, cleaved form; Mpf; Msln; Pre-pro-megakaryocyte-potentiating factor; SMR; SMRP; soluble MPF mesothelin related protein | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.