Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesothelin Antibody (MSLN/2131) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279858
Description
Mesothelin Monoclonal specifically detects Mesothelin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Mesothelin | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG2b κ |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
MSLN/2131 | |
Unconjugated | |
10 mM PBS with No Preservative | |
MSLN | |
Recombinant fragment (around aa 273-407) of human Mesothelin (MSLN) protein (exact sequence is proprietary) (Uniprot: Q13421) | |
100 μg | |
Cancer | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction