Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesothelin (MSLN) Monoclonal Antibody (OTI7C2), TrueMAB™, OriGene
SDP

Catalog No. 502543405
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3405 Supplier OriGene Supplier No. CF814168
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mesothelin (MSLN) Monoclonal antibody specifically detects Mesothelin (MSLN) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Mesothelin (MSLN)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI7C2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene FABP3
Gene Accession No. P05413
Gene Alias CAK1; CAK1 antigen; megakaryocyte potentiating factor; Megakaryocyte-potentiating factor; Mes; Mesothelin; Mesothelin, cleaved form; MPF; MSLN; Pre-pro-megakaryocyte-potentiating factor; SMR; SMRP; soluble MPF mesothelin related protein
Host Species Mouse
Immunogen Full length human recombinant protein of human FABP3 (NP_004093) produced in EXPI-293F cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2170
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.