Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mesothelin Recombinant Rabbit Monoclonal Antibody (1I1Y7)
GREENER_CHOICE

Catalog No. FEMA551345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551345 Supplier Invitrogen™ Supplier No. MA551345
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: EVEKTACPSG KKAREIDESL IFYKKWELEA CVDAALLATQ MDRVNAIPFT YEQLDVLKHK LDELYPQGYP ESVIQHLGYL FLKMSPEDIR KWNVTSLETL KALLEVNKGH EMSPQAPRRP LPQVATLIDR FVKGRGQLDK DTLDTLTAFY PGYLCSLSPE ELSSVPPSSI WAVRPQDLDT CDPRQLDVLY PKARLAFQNM NGSEYFVKIQ SFLGGAPTED LKALSQQNVS MDLATFMKLR TDAVLPLTVA EVQKLLGPHV EGLKAEERHR PVRDWILRQR QDDLDTLGLG LQGGIPNGYL VLDLSMQEAL S.

Originally identified as a differentiation antigen of mesotheliomas, ovarian cystadenocarcinomas, and pancreatic adenocarcinomas, Mesothelin is a glycosylphosphatidylinositol (GPI)-anchored, cell-surface glycoprotein predominantly secreted by cells of the mesothelium. Although Mesothelin is expressed at restricted levels by normal mesothelial cells of the pleural,pericardial, and peritoneal membranes, aberrant expression has been documented in the aforementioned cancers, as well as in endometriod uterine adenocarcinomas and squamous cell carcinomas of the esophagus, stomach, lung, and cervix. Proteolytic cleavage of Mesothelin yields a soluble, polypeptide fragment designated megakaryocyte potentiating factor (MPF) based on its ability to stimulate megakaryocyte colony-forming activity of murine interleukin-3 in murine bone marrow cell cultures. Originally isolated from the HPC-Y5 pancreatic cell line, MPF has been suggested to play a role in the proliferation and differentiation of megakaryoctyes, and the regulation of resultant platelet production. While the biological functions of both Mesothelin and MPF remain speculative, high-levels of expression in cancerous tissues compared to limited distribution in normal tissues strongly suggests their involvement in tumorigenesis. Both have been demonstrated to promote tumor cell proliferation, migration,anchorage-independent growth, and tumor progression; demonstrating their involvement in heterotypic cell adhesion and the metastatic spread of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mesothelin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1I1Y7
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MSLN
Gene Accession No. Q13421
Gene Alias CAK1; CAK1 antigen; megakaryocyte potentiating factor; Megakaryocyte-potentiating factor; Mes; Mesothelin; Mesothelin, cleaved form; MPF; MSLN; Pre-pro-megakaryocyte-potentiating factor; SMR; SMRP; soluble MPF mesothelin related protein
Gene Symbols MSLN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 296-606 of human Mesothelin (NP_037536.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10232
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.