Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesothelioma Antibody (ME1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222442
Description
Mesothelioma Monoclonal specifically detects Mesothelioma in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Radioimmunoassay, Western Blot (Negative).Specifications
Mesothelioma | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 2 - 5 μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 2 - 5 μg/mL, Radioimmunoassay 1:100 - 1:2000, Western Blot (Negative) | |
MMS | |
Mesothelioma cell line SPC111. | |
25 μg | |
Primary | |
This antibody reacts with Human. | |
Human | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot | |
ME1 | |
Unconjugated | |
Malignant mesothelioma, susceptibility to | |
Mouse | |
Protein A purified | |
RUO | |
338340 | |
Reconstitute with PBS. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction