Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesothelioma Monoclonal Antibody (HBME-1), Invitrogen™

Catalog No. PIMA512220
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512220 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512220 Supplier Thermo Scientific Supplier No. MA512220
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-12220 targets Mesothelioma in IHC (P) applications and shows reactivity with Human samples. The MA5-12220 immunogen is a suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma.

Mesothelioma Monoclonal antibody specifically detects Mesothelioma in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mesothelioma
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HBME-1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Host Species Mouse
Immunogen A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.