Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesp1 Antibody (2030B), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB9219S
Description
Mesp1 Monoclonal antibody specifically detects Mesp1 in Human samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
Mesp1 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
2030B | |
Western Blot, Immunocytochemistry | |
BHLHC5, bHLHc5MGC10676, Class C basic helix-loop-helix protein 5, mesoderm posterior 1 homolog (mouse), mesoderm posterior protein 1 | |
E. coli-dervived recombinant human MESP1, Met1-Gln85, Accession # Q9BRJ9 | |
0.1 mL | |
Cardiovascular Biology, Developmental Biology, Stem Cells | |
55897 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction