Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MESP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587675
Description
Immunogen sequence: RRCRQRGDAG SPRGCPLCPD DCPAQMQTRT QAEGQGQGRG LGLVSAVRAG ASWGSPPACP GARAAPEPRD PPALFAEAAC PEGQAMEPSP PSPLLPGDVL ALLETWMPLS PLEWLPEEPK; Positive Samples: A-549, HeLa, THP-1, Mouse heart, Rat heart; Cellular Location: Nucleus.
Mesp1 (mesoderm posterior 1) is a basic helix-loop-helix-type transcription factor, which along with its related proteins, has an essential role in both segmentation and rostro-caudal patterning of somites in the anterior pre-somitic mesoderm. Mesp1 is expressed in many of the precursors of the cardiovascular system and is known to play a role in the process of cardiac morphogenesis.
Specifications
MESP1 | |
Polyclonal | |
Unconjugated | |
MESP1 | |
bHLHc5; Class C basic helix-loop-helix protein 5; mesoderm posterior 1; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; mesoderm posterior bHLH transcription factor 1; mesoderm posterior protein 1; MESP1; MGC10676 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
17292, 308766, 55897 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97309, Q9BRJ9 | |
MESP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 149-268 of human MESP1 (NP_0611401). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction