Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MESP2 Antibody (1B3F9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | MESP2 |
---|---|
Clone | 1B3F9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Description
MESP2 Monoclonal specifically detects MESP2 in Human samples. It is validated for Western Blot, ELISA.Specifications
MESP2 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.05% Sodium Azide | |
145873 | |
Purified recombinant fragment of human MESP2 (AA: 37-94 ) expressed in E. Coli. | |
Primary | |
41.8 kDa |
1B3F9 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Mouse | |
Human | |
bHLHc6, class C basic helix-loop-helix protein 6, mesoderm posterior 2 homolog (mouse), mesoderm posterior protein 2, SCDO2 | |
MESP2 | |
IgG1 | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title