Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MESP2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen MESP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP18021920
SDP
View Documents
Novus Biologicals
NBP18021920UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP180219
SDP
View Documents
Novus Biologicals
NBP180219
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

MESP2 Polyclonal specifically detects MESP2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

MESP2
Polyclonal
Purified
RUO
bHLHc6, class C basic helix-loop-helix protein 6, mesoderm posterior 2 homolog (mouse), mesoderm posterior protein 2, SCDO2
MESP2
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
NP_032615
145873
Synthetic peptide directed towards the C terminal of mouse MESP2. Peptide sequence SCQRLQPQPQPQPQWGCWGHDAEVLSTSEDQGSSPALQLPVASPTPSSGL.
Primary
41 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.