Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MESP2 Monoclonal Antibody (1B3F9)
GREENER_CHOICE

Catalog No. PIMA517121
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517121 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517121 Supplier Invitrogen™ Supplier No. MA517121
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17121 targets MESP2 in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17121 immunogen is purified recombinant fragment of human MESP2 (amino acids: 37-94 ) expressed in E. Coli. MA5-17121 detects MESP2 which has a predicted molecular weight of approximately 41.8kDa.

This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MESP2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B3F9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Mesp2
Gene Accession No. Q0VG99
Gene Alias bHLHc6; Class C basic helix-loop-helix protein 6; mesoderm posterior 2; mesoderm posterior 2 homolog; mesoderm posterior basic helix-loop-helix transcription factor 2; mesoderm posterior bHLH transcription factor 2; mesoderm posterior protein 2; Mesp2; SCDO2
Gene Symbols Mesp2
Host Species Mouse
Immunogen Purified recombinant fragment of human MESP2 (amino acids: 37-94 ) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145873
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.