Learn More
Invitrogen™ MESP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA5143961
Catalog No. PIPA5143961
Only null left
Add to cart
Description
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human A549 whole cell, human SW620 whole cell. IHC: human lung cancer tissue. ICC/IF: A549 cell. Flow: K562 cell, SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MESP2
Polyclonal
Unconjugated
Mesp2
bHLHc6; Class C basic helix-loop-helix protein 6; mesoderm posterior 2; mesoderm posterior 2 homolog; mesoderm posterior basic helix-loop-helix transcription factor 2; mesoderm posterior bHLH transcription factor 2; mesoderm posterior protein 2; Mesp2; SCDO2
Rabbit
Antigen Affinity Chromatography
RUO
145873
-20°C
Lyophilized
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
500 μg/mL
PBS with 4mg trehalose and 0.01mg sodium azide
Q0VG99
Mesp2
E.coli-derived human Mesp2 recombinant protein (Position: M1-Y397).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.