Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Met Enkephalin Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1759R

Description
Methionine enkephalin (met enkephalin) is a pentapeptide that appears to enhance immune function at low doses and suppresse at high doses. This molecule acts in the central nervous, neuroendocrine, and immune systems. Cells from these systems have receptors for met enkephalin and are able to generate met enkephalin from its prohormone, proenkephalin A. Met enkephalin may be capable of enhancing immune function in cancer or AIDS patients.Specifications
Met Enkephalin | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
KLH conjugated Met Enkephalin peptide (YGGFM). | |
100 μL | |
Primary | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction