Learn More
Promega MET (M1250T) Kinase Enzyme System, 10microg
Catalog No. PRV4168 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4168 10 μg
1 options
Catalog No. PRV4168 Supplier Promega Supplier No. V4168
Add to Cart
Add to Cart
Recombinant human MET (point mutant M1250T; amino acids 956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile MET (M1250T) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C