Learn More
Promega MET (M1250T) Kinase Enzyme System, 1mg
Catalog No. PRV6448 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6448 1 mg
1 options
Catalog No. PRV6448 Supplier Promega Supplier No. V6448
Add to Cart
Add to Cart
Request a Quote
Recombinant human MET (point mutant M1250T; amino acids 956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile MET (M1250T) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C