Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meta-Pneumovirus Antibody (HMPV123) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP121631
Description
Meta-Pneumovirus Monoclonal specifically detects Meta-Pneumovirus in Virus samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Meta-Pneumovirus | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:40-1:80, Immunoprecipitation | |
| GLYG | |
| Sub-type A hMPV virus isolate NCL03-4/145 | |
| 0.1 mg | |
| Primary | |
| NBP1-21631 recognises the nucleoprotein of all sub-types (A1, A2, B1 and B2) of hMPV. NBP1-21631 does not cross-react with cell cultures infected with hRSV, influenzaviruses A and B, adenovirus, parainfluenza viruses 1, 2, 3 and 4b, mumps virus, measles virus, varicella-zoster virus, herpes simplex virus types 1 and 2, human cytomegalovirus, human herpesvirus type 6, ECHOvirus 19, Coxsachievirus B4, poliovirus types 1-3, HHV6 or uninfected HeLa, MA104, 3MK, LLC-MK, HEp2, MRC-5 and HSB-2 cells. It also does not react with sputum bacteria. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| HMPV123 | |
| Unconjugated | |
| hMPV, metapneumovirus | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 2799942 | |
| Primate, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction