Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meta-Pneumovirus Antibody (HMPV123), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP121631PECY7
Description
Meta-Pneumovirus Monoclonal specifically detects Meta-Pneumovirus in Virus samples. It is validated for Flow Cytometry.Specifications
Meta-Pneumovirus | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
GLYG | |
Sub-type A hMPV virus isolate NCL03-4/145 | |
0.1 mL | |
2799942.0 | |
Virus | |
IgG2a |
Flow Cytometry | |
HMPV123 | |
Flow Cytometry | |
hMPV, metapneumovirus | |
Mouse | |
Protein G purified | |
Primary | |
NBP1-21631 recognises the nucleoprotein of all sub-types (A1, A2, B1 and B2) of hMPV. NBP1-21631 does not cross-react with cell cultures infected with hRSV, influenzaviruses A and B, adenovirus, parainfluenza viruses 1, 2, 3 and 4b, mumps virus, measles virus, varicella-zoster virus, herpes simplex virus types 1 and 2, human cytomegalovirus, human herpesvirus type 6, ECHOvirus 19, Coxsachievirus B4, poliovirus types 1-3, HHV6 or uninfected HeLa, MA104, 3MK, LLC-MK, HEp2, MRC-5 and HSB-2 cells. It also does not react with sputum bacteria. | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction