Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metadherin Antibody (OTI1B6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272552PE
Description
Metadherin Monoclonal antibody specifically detects Metadherin in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Metadherin | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
OTI1B6 | |
Western Blot, Immunohistochemistry | |
3D3, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, protein LYRIC | |
Human recombinant protein fragment corresponding to amino acids 70-582 of human Metadherin (NP_848927)produced in SF9 cell. | |
0.1 mL | |
Phospho Specific | |
92140 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction