Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Metadherin Monoclonal Antibody (CL0397)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA534982
Description
Immunogen sequence: RKTEPSAWSQ DTGDANTNGK DWGRSWSDRS IFSGIGSTAE PVSQSTTSDY QWDVSRNQPY IDDEWSGLNG LSSADPNSDW NAPAEEWGNW VDEERASLLK SQEPIPDDQK VSDDDKEKGE GALPTGKSKK KKKKKKKQGE DNSTAQD.
Metadherin (Metastasis adhesion protein), also known as MTDH, LYsine-RIch CEACAM1 co-isolated (LYRIC), is a novel protein that localizes with the tight junction proteins ZO-1 and occludin in polarized epithelial cells. At the tight junction, it acts not as a structural component, but is rather recruited during the maturation of the tight junction complex. Metadherin is overexpressed in breast cancer tissue and breast tumor xenografts, while much lower levels are expressed in normal breast tissue. Metadherin binds to lung vasculature, one of the four common sites of breast cancer metastasis, through a C-terminal segment in the extracellular domain; blocking this lung-homing domain with antibodies or inhibiting metadherin with siRNA has been reported to inhibit breast cancer metastasis.
Specifications
Metadherin | |
Monoclonal | |
Unconjugated | |
MTDH | |
2610103J23Rik; 3D3; 3D3/Lyric; AEG1; AEG-1; astrocyte elevated gene 1; astrocyte elevated gene-1 protein; AV353288; D8Bwg1112e; LYRIC; LYRIC/ MTDH; LYRIC/3D3; lysine-rich CEACAM1 co-isolated protein; Metadherin; metastasis adhesion protein; MTDH; Protein LYRIC | |
Mouse | |
Protein A | |
RUO | |
92140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
CL0397 | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UE4 | |
MTDH | |
Recombinant Protein corresonding to Human Metadherin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction