Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Metal Retort Base Stand

This stand base is made of a lightweight, yet solid metal. An epoxy coating makes it chemically resistant. The stand contains a threaded opening at one end 6/16 in. in diameter.
Supplier: Eisco™ CH0649B
Description
- Rectangular retort base made of corrosion resistant, powder coated pressed steel - Features rubber strips to protect surfaces
- Base features a tapped hole for a 10 mm x 1.5 mm threaded rod (sold separately). Best used with Eisco Labs rod series: CH0657, CH0658, CH0659
- The durable and sturdy construction of this set makes it an excellent choice for science classrooms and basic professional laboratory setups
Specifications
Metal | |
1 in. | |
20 cm | |
13 cm | |
Retort Base Stand | |
Epoxy | |
1.22 kg |
2.54 cm | |
8 in. | |
5 in. | |
Threaded opening to hold equipment approximately 0.5 in. diameter, Rubber strips on feet to protect standing surface | |
Blue | |
2.7 lb. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction