Learn More
Eisco™ Metal Retort Base Stand
This stand base is made of a lightweight, yet solid metal. An epoxy coating makes it chemically resistant. The stand contains a threaded opening at one end 6/16 in. in diameter.
$36.85 - $63.60
Description
- Rectangular retort base made of corrosion resistant, powder coated pressed steel - Features rubber strips to protect surfaces
- Base features a tapped hole for a 10 mm x 1.5 mm threaded rod (sold separately). Best used with Eisco Labs rod series: CH0657, CH0658, CH0659
- The durable and sturdy construction of this set makes it an excellent choice for science classrooms and basic professional laboratory setups
Specifications
Retort Base Stand | |
Blue |