Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Metallothionein Monoclonal Antibody (2B5), FITC
GREENER_CHOICE

Catalog No. PIMA545899
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545899 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545899 Supplier Invitrogen™ Supplier No. MA545899
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45899 was sufficient for detection of Metallothionein in 1 μg of purified Pseudomonas aeruginosa Metallothionein (PmtA)-GST tagged protein by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects 36 kDa band corresponding to Pseudomonas aeruginosa Metallothionein.

Metallothioneins (MTs) are a family of low molecular weight, heavy metal-binding proteins characterized by a high cysteine content and lack of aromatic amino acids. MTs bind 7 to 12 heavy metal atoms per molecule of protein. They are ubiquitous in the animal and plant kingdoms and are also found in prokaryotes. In mammals, the cysteine residues are absolutely conserved and serve to coordinate heavy metal atoms such as zinc, cadmium, and copper via mercaptide linkages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Metallothionein
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B5
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene Mt1
Gene Accession No. 0
Gene Alias CES 1; CES1; LOC100343802; LOC100630794; Metallothionein; metallothionein 1; metallothionein 1a; metallothionein 1A (functional); metallothionein 1S; Metallothionein 2; Metallothionein 2A; Metallothionein IA; Metallothionein II; metallothionein-1; Metallothionein-1A; metallothionein-1A-like; Metallothionein2; metallothionein-I; Metallothionein-IA; MGC32848; Mt; MT 1A; MT 2; MT 2A; MT IA; MT II; Mt1; MT-1; MT1A; MT-1A; MT1S; MT2; MT2A; MTC; MT-I; MT-IA; Probable pmtA
Gene Symbols Mt1, PA2140
Host Species Mouse
Immunogen Recombinant full length Pseudomonas aeruginosa Metallothionein protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20220066, 880550
Target Species Bacteria
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.