Learn More
Invitrogen™ Metallothionein Monoclonal Antibody (8D8), FITC

Description
A 1:1,000 dilution of MA5-45903 was sufficient for detection of Metallothionein in 1 μg of purified Pseudomonas aeruginosa Metallothionein (PmtA)-GST tagged protein by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects 36 kDa band corresponding to Pseudomonas aeruginosa Metallothionein.
Specifications
Specifications
| Antigen | Metallothionein |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 8D8 |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 |
| Gene | Mt1 |
| Gene Accession No. | 0 |
| Gene Alias | CES 1; CES1; LOC100343802; LOC100630794; Metallothionein; metallothionein 1; metallothionein 1a; metallothionein 1A (functional); metallothionein 1S; Metallothionein 2; Metallothionein 2A; Metallothionein IA; Metallothionein II; metallothionein-1; Metallothionein-1A; metallothionein-1A-like; Metallothionein2; metallothionein-I; Metallothionein-IA; MGC32848; Mt; MT 1A; MT 2; MT 2A; MT IA; MT II; Mt1; MT-1; MT1A; MT-1A; MT1S; MT2; MT2A; MTC; MT-I; MT-IA; Probable pmtA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.