Learn More
Eisco™ Metamorphic Rocks Kit
SCI_ED
Catalog No. S24666 Shop All Eisco Products
Change view
Click to view available options
Quantity:
12 Specimens
Catalog No. Quantity
S24666 12 Specimens
1 options
Catalog No. S24666 Supplier Eisco™ Supplier No. ESNG0007
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For study of foliation and alteration of these metamorphic rocks to understand the immense heat and pressure needed to form them. The Metamorphic Rocks Kit contains 12 specimens measuring approx. 3 cm (1 in.) and specimen key.
  • Classroom tool for metamorphic rock classification and identification
  • Numbered and stored in sturdy, sectioned container for easy access and safety
  • Average specimen size: 2 to 3 cm
  • Classified based on whether or not they are foliated, as well as grade of metamorphism
Specifications
Product Type Metamorphic Rock Kit
For Use With (Application) Study foliation/alteration to understand immense heat and pressure needed to form them
Includes Anthracite coal, banded gneiss, augen gneiss, fine and coarse marble, phyllite, white and pink quartzite, mica schist, garnet schist, serpentinite, slate
Packaging Sectioned container
Quantity 12 Specimens
Specimen Size Approx. 1 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov