Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Melting Point | 86°C |
| Color | Lilac |
| Physical Form | Fine Crystalline Powder |
| Quantity | 100 g |
| Assay Percent Range | 98.0 to 102% (Complexometry) |
| Linear Formula | CoCl2·6H2O |
| Fieser | 01,155; 06,127; 11,479 |
| Merck Index | 15, 2421 |
| Solubility Information | Solubility in water: 970g/L (20°C) |
| Formula Weight | 237.93 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May damage fertility. Harmful if inhaled. Harmful if swallowed. Suspected of causing genetic defects. May cause cancer by inhalation. May cause an allergic skin reaction. May cause allergy or ast |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: |
| RTECSNumber | GG0200000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction