Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544285 Supplier Invitrogen™ Supplier No. PA544285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GDINTDPWAG YRYTGKLRPH YPLMPTRPVP SYIQRPDYAD HPLGMSESEQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 82%.

Methionine aminopeptidase 1 (METAP1) is a gene located on chromosome 4q24, encoding an enzyme responsible for the removal of N-terminal methionine residues from newly synthesized proteins. METAP1 is essential for protein maturation and function, as the cleavage of methionine allows subsequent modifications that are crucial for protein stability and activity. This enzyme is ubiquitously expressed in various tissues and plays a significant role in post-translational modification processes, affecting cellular homeostasis and protein regulation. METAP1 is involved in several physiological pathways, including cell cycle progression, signal transduction, and stress response. Dysregulation of METAP1 activity can impact protein function and is associated with diseases like cancer, reflecting its importance in cellular proliferation and growth. Research into METAP1 focuses on its potential as a therapeutic target for modulating protein synthesis and its implications in diseases characterized by uncontrolled cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METAP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene METAP1
Gene Accession No. P53582
Gene Alias 1700029C17Rik; AW047992; DKFZp781C0419; EC 3.4.11.18; fc84e12; im:7047238; KIAA0094; KIAA0094MAP 1; MAP 1; MAP1; MAP1A; metAP 1; Metap1; metap1 protein; MetAP1A; Methionine aminopeptidase 1; methionyl aminopeptidase 1; mKIAA0094; Peptidase M; Peptidase M 1; Putative methionine aminopeptidase 1 (MetAP 1) (Peptidase M 1); wu:fc84e12; zgc:110093
Gene Symbols METAP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human METAP1 (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23173
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.