Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METAP1D Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547655
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The N-terminal methionine excision pathway is an essential process in which the N-terminal methionine is removed from many proteins, thus facilitating subsequent protein modification. In mitochondria, enzymes that catalyze this reaction are celled methionine aminopeptidases (MetAps, or MAPs; EC 3.4.11.18).
Specifications
| METAP1D | |
| Polyclonal | |
| Unconjugated | |
| METAP1D | |
| 2310066F24Rik; 3110033D18Rik; AV117938; CDS of metAP-3 within PCR fragment; MAP 1D; Map1d; MetAP 1D; METAP1D; Metap1l; Metapl1; methionine aminopeptidase 1D, mitochondrial; methionine aminopeptidase-like 1; methionyl aminopeptidase type 1D (mitochondrial); methionyl aminopeptidase type 1D, mitochondrial; Mnpepl; Peptidase M 1D | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 254042 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q6UB28 | |
| METAP1D | |
| Sf 21-derived recombinant human Methionine Aminopeptidase 1D/MAP1D Arg44-Ala335. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction