Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METAP1D Recombinant Rabbit Monoclonal Antibody (008), Invitrogen™

Catalog No. PIMA543434
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543434 50 μL
PIMA543126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543434 Supplier Invitrogen™ Supplier No. MA543434
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human MAP1D

The N-terminal methionine excision pathway is an essential process in which the N-terminal methionine is removed from many proteins, thus facilitating subsequent protein modification. In mitochondria, enzymes that catalyze this reaction are celled methionine aminopeptidases (MetAps, or MAPs; EC 3.4.11.18).
TRUSTED_SUSTAINABILITY

Specifications

Antigen METAP1D
Applications ELISA
Classification Recombinant Monoclonal
Clone 008
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene METAP1D
Gene Accession No. Q6UB28
Gene Alias 2310066F24Rik; 3110033D18Rik; AV117938; CDS of metAP-3 within PCR fragment; MAP 1D; Map1d; MetAP 1D; METAP1D; Metap1l; Metapl1; methionine aminopeptidase 1D, mitochondrial; methionine aminopeptidase-like 1; methionyl aminopeptidase type 1D (mitochondrial); methionyl aminopeptidase type 1D, mitochondrial; Mnpepl; Peptidase M 1D
Gene Symbols METAP1D
Host Species Rabbit
Immunogen Recombinant Human MAP1D protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254042
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.