Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metapneumovirus Antibody (hMPV57) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $501.50
Specifications
Antigen | Metapneumovirus |
---|---|
Clone | hMPV57 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Description
Metapneumovirus Monoclonal specifically detects Metapneumovirus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Metapneumovirus | |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Virology Bacteria and Parasites | |
PBS with 0.02% Sodium Azide | |
IgG1 | |
Protein A purified |
hMPV57 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
Human MPV strain NCL-174 | |
Primary | |
Identifies Metapneumovirus nucleoprotein of both types A and B. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title