Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metapneumovirus Fusion Protein Antibody (hMPV24), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250505G
Description
Metapneumovirus Fusion Protein Monoclonal specifically detects Metapneumovirus Fusion Protein in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Metapneumovirus Fusion Protein | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Human MPV strain NCL-145 | |
0.1 mL | |
Virology Bacteria and Parasites | |
Virus | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
hMPV24 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction