Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Metapneumovirus matrix Monoclonal Antibody (J0J)
GREENER_CHOICE

Catalog No. PIMA518223
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518223 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518223 Supplier Invitrogen™ Supplier No. MA518223
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific for the matrix protein of human metapneumovirus, hMPV genotypes A & B. It does not cross-react with Respiratory Syncytial Virus, Influenza A, Influenza B, Adenovirus or Parainfluenza viruses 1, 2 & 3. Purity is >90%. Recommended pairs for lateral flow: MA5-18222 (capture) with MA5-18223 (conjugated, detection).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Metapneumovirus matrix
Applications ELISA, Lateral Flow, Immunocytochemistry
Classification Monoclonal
Clone J0J
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias hMPV; h-MPV; hMPV matrix; MPV matrix protein
Host Species Mouse
Immunogen Metapneumovirus recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.