Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Metapneumovirus Monoclonal Antibody (HMPV33)
GREENER_CHOICE

Catalog No. PIMA516714
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516714 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516714 Supplier Invitrogen™ Supplier No. MA516714
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Metapneumovirus antibody, clone HMPV33 recognizes human metapneumovirus (hMPV), a member of the Paramyxoviridae RNA virus family.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Metapneumovirus
Applications Immunocytochemistry
Classification Monoclonal
Clone HMPV33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias h h h-MPV; MPV
Host Species Mouse
Immunogen Sub-type A hMPV virus isolate NCL03-4/174.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.