Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metapneumovirus Phosphoprotein Antibody (hMPV33), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250506C
Description
Metapneumovirus Phosphoprotein Monoclonal specifically detects Metapneumovirus Phosphoprotein in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Metapneumovirus Phosphoprotein | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Human hMPV strain NCL-174 | |
0.1 mL | |
Virology Bacteria and Parasites | |
Identifies Metapneumovirus nucleoprotein of both types A and B. | |
Store at 4C in the dark. | |
IgG2a |
ELISA, Immunocytochemistry, Immunofluorescence | |
hMPV33 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction