Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MetaSystems CL ATM (11q22) DNA Probe
SDP

Catalog No. 22200358
Encompass
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-200-358 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-200-358 Supplier MetaSystems Supplier No. D5992100OR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Includes

Orange labeled probe

Detects deletions which may occur in long arm of chromosome 11 including ATM gene

  • Orange labeled probe hybridizes to area spanning ATM gene and NPAT at 11q22

Specifications

Concentration 100 μL
Content And Storage Orange labeled probe
Quantity 10 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.