Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MetaSystems CL TP53 DNA Probe
SDP

Catalog No. 22200356
Encompass
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-200-356 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-200-356 Supplier MetaSystems Supplier No. D5988100OR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Includes

Orange labeled probe

Detects deletions which may occur in short arm of chromosome 17 including TP53 gene

  • Orange labeled probe hybridizes to area spanning TP53 at 17p13

Specifications

Concentration 100 μL
Content And Storage Orange labeled probe
Quantity 10 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.